2-(Methoxycarbonyl)nicotinic acid 96%

Lieferant: Apollo Scientific

OR901680-1G OR901680-5G
APOSOR901680-1GEA 81.2 EUR
APOSOR901680-1G APOSOR901680-5G
2-(Methoxycarbonyl)nicotinic acid 96%
2-(Methoxycarbonyl)nicotinic acid
Formel: C₈H₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00111560
CAS-Nummer: 24195-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR