6-Brom-2H-1,4-benzoxazin-3(4H)-on

Lieferant: Apollo Scientific
Achtung

OR901681-25G
APOSOR901681-25GEA 111 EUR
APOSOR901681-25G
6-Brom-2H-1,4-benzoxazin-3(4H)-on
6-Bromo-2H-1,4-benzoxazin-3(4H)-one
Formel: C₈H₆BrNO₂
Molecular Weight: 228,04 g/mol
Schmelzpunkt: 220…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00461173
CAS-Nummer: 24036-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR