6-Phenyl-2-pyridone 98%

Lieferant: Apollo Scientific

Synonyme: 6-Phenylpyridin-2(1H)-one

OR901687-1G OR901687-5G
APOSOR901687-1GEA 219 EUR
APOSOR901687-1G APOSOR901687-5G
6-Phenyl-2-pyridone 98%
6-Phenyl-2-pyridone
Formel: C₁₁H₉NO
Molecular Weight: 171,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059760
CAS-Nummer: 19006-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR