2-Methoxy-6,8-dimethylchinolin 98%

Lieferant: Apollo Scientific

OR901693-1G OR901693-25G OR901693-5G
APOSOR901693-1GEA 145 EUR
APOSOR901693-1G APOSOR901693-25G APOSOR901693-5G
2-Methoxy-6,8-dimethylchinolin 98%
2-Methoxy-6,8-dimethylchinolin
Formel: C₁₂H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD15143628
CAS-Nummer: 861581-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR