3,5-Dichlorpyridin-N-oxid 98%
Lieferant: Apollo Scientific
Synonyme:
3,5-Dichloropyridine1-oxide
OR901695-25G
APOSOR901695-25GEA
259
EUR
APOSOR901695-25G
3,5-Dichlorpyridin-N-oxid 98%
3,5-Dichlorpyridin-N-oxid
Formel:
C₅H₃Cl₂NO Molecular Weight: 163,9894 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00955620 CAS-Nummer: 15177-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...