2-Ethoxy-8-methylchinolin 98%
Lieferant: Apollo Scientific
OR901702-100G
OR901702-25G
OR901702-5G
APOSOR901702-100GEA
632
EUR
APOSOR901702-100G
APOSOR901702-25G
APOSOR901702-5G
2-Ethoxy-8-methylchinolin 98%
2-Ethoxy-8-methylquinoline
Formel:
C₁₂H₁₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD16036158 CAS-Nummer: 1221793-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...