5-Bromo-N-ethyl-2-pyridinamine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-ethylaminopyridine
OR901754-10G
OR901754-1G
OR901754-5G
APOSOR901754-10GEA
1230
EUR
APOSOR901754-10G
APOSOR901754-1G
APOSOR901754-5G
5-Bromo-N-ethyl-2-pyridinamine 98%
5-Bromo-N-ethyl-2-pyridinamine
Formel:
C₇H₉BrN₂ Molecular Weight: 201,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09743642 CAS-Nummer: 856850-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...