2-Bromo-4-hydroxypyridine 96%

Lieferant: Apollo Scientific

Synonyme: 2-Bromopyridin-4-ol

OR901771-10G OR901771-1G OR901771-5G
APOSOR901771-10GEA 255 EUR
APOSOR901771-10G APOSOR901771-1G APOSOR901771-5G
2-Bromo-4-hydroxypyridine 96%
2-Bromo-4-hydroxypyridine
Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
Schmelzpunkt: 171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234040
CAS-Nummer: 36953-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR