3-Amino-5-chlor-2-methylpyridin 98%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-5-chloropicoline, 5-Chloro-2-methylpyridin-3-amine

OR901812-1G OR901812-5G
APOSOR901812-1GEA 121 EUR
APOSOR901812-1G APOSOR901812-5G
3-Amino-5-chlor-2-methylpyridin 98%
3-Amino-5-chloro-2-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142.5862 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12196974
CAS-Nummer: 89639-36-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR