2,4-Dimethoxypyrimidin 98%

Lieferant: Apollo Scientific

OR901864-100G
APOSOR901864-100GEA 172 EUR
APOSOR901864-100G
2,4-Dimethoxypyrimidin 98%
2,4-Dimethoxypyrimidine
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00056028
CAS-Nummer: 3551-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR