2-(Methylthio)pyridin 95%

Lieferant: Apollo Scientific

OR901942-25G OR901942-5G
APOSOR901942-25GEA 0 EUR
APOSOR901942-25G APOSOR901942-5G
2-(Methylthio)pyridin 95%
2-(Methylthio)pyridine
Formel: C₆H₇NS
Lagertemperatur: Raumtemperatur
MDL: MFCD01691593
CAS-Nummer: 18438-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR