6,7-Dichlorchinolin 98%

Lieferant: Apollo Scientific

OR901961-1G OR901961-5G
APOSOR901961-1GEA 168 EUR
APOSOR901961-1G APOSOR901961-5G
6,7-Dichlorchinolin 98%
6,7-Dichloroquinoline
Formel: C₉H₅Cl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD09878431
CAS-Nummer: 40635-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR