4-Methoxynicotinsäure 98%

Lieferant: Apollo Scientific

OR901967-1G OR901967-5G
APOSOR901967-1GEA 56.7 EUR
APOSOR901967-1G APOSOR901967-5G
4-Methoxynicotinsäure 98%
4-Methoxynicotinsäure
Formel: C₇H₇NO₃
Molecular Weight: 153,1400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115724
CAS-Nummer: 10177-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR