3-Bromo-6-mercaptopyridine 98%

Lieferant: Apollo Scientific

Synonyme: 5-Bromopyridine-2-thiol

OR902013-5G
APOSOR902013-5GEA 78.3 EUR
APOSOR902013-5G
3-Bromo-6-mercaptopyridine 98%
3-Bromo-6-mercaptopyridine
Formel: C5H4BrNS
Molecular Weight: 190,0600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09729468
CAS-Nummer: 56673-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR