2-Brom-4-methylpyrimidin 97%

Lieferant: Apollo Scientific

OR902029-1G OR902029-25G OR902029-5G
APOSOR902029-1GEA 74.1 EUR
APOSOR902029-1G APOSOR902029-25G APOSOR902029-5G
2-Brom-4-methylpyrimidin 97%
2-Bromo-4-methylpyrimidine
Formel: C₅H₅ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04037883
CAS-Nummer: 130645-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR