2-(Boc-amino)-6-chlorpyridin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (6-chloropyridin-2-yl)carbamate, 2-Amino-6-chloropyridine, 2-BOC protected

OR902051-1G OR902051-25G OR902051-5G
APOSOR902051-1GEA 77 EUR
APOSOR902051-1G APOSOR902051-25G APOSOR902051-5G
2-(Boc-amino)-6-chlorpyridin 97%
2-(Boc-amino)-6-chlorpyridin
Formel: C₁₀H₁₃ClN₂O₂
Molecular Weight: 228,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09030821
CAS-Nummer: 159603-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR