2-Methoxypyrimidin 98%

Lieferant: Apollo Scientific

OR902052-1G OR902052-5G
APOSOR902052-1GEA 207 EUR
APOSOR902052-1G APOSOR902052-5G
2-Methoxypyrimidin 98%
2-Methoxypyrimidine
Formel: C₅H₆N₂O
Siedepunkt: 69…70 °C/22mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00234104
CAS-Nummer: 931-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR