3-Bromo-4-hydroxypyridin-2(1H)-one 95%
Lieferant: Apollo Scientific
Synonyme:
3-Bromopyridine-2,4-diol
OR902060-10G
OR902060-1G
OR902060-5G
APOSOR902060-10GEA
928
EUR
APOSOR902060-10G
APOSOR902060-1G
APOSOR902060-5G
3-Bromo-4-hydroxypyridin-2(1H)-one 95%
3-Bromo-4-hydroxypyridin-2(1H)-one
Formel:
C5H4BrNO2 Molecular Weight: 190 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11101040 CAS-Nummer: 96245-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...