2-Chlor-N-methylisonicotinamid 98%

Lieferant: Apollo Scientific

Synonyme: N-Methyl 2-chloroisonicotinamide

OR902089-1G OR902089-5G
APOSOR902089-1GEA 95.7 EUR
APOSOR902089-1G APOSOR902089-5G
2-Chlor-N-methylisonicotinamid 98%
2-Chlor-N-methylisonicotinamid
Formel: C₇H₇ClN₂O
Molecular Weight: 170,6000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09862946
CAS-Nummer: 131418-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR