6-Chlorchinolin-2(1H)-on 98%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-2-hydroxyquinoline

OR902094-5G
APOSOR902094-5GEA 37.7 EUR
APOSOR902094-5G
6-Chlorchinolin-2(1H)-on 98%
6-Chlorchinolin-2(1H)-on
Formel: C₉H₆ClNO
Molecular Weight: 179,6030 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02930633
CAS-Nummer: 1810-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR