2,4-Dimethylphenylhydraziniumchlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dimethylphenylhydrazin Hydrochlorid, 1-(2,4-Dimethylphenyl)hydrazin Hydrochlorid, 2,4-Dimethylphenylhydrazinhydrochlorid

OR9021-100G OR9021-25G OR9021-10G
APOSOR9021-100GEA 168 EUR
APOSOR9021-100G APOSOR9021-25G APOSOR9021-10G
2,4-Dimethylphenylhydraziniumchlorid 97%
2,4-Dimethylphenylhydraziniumchlorid
Formel: C₈H₁₃ClN₂
Molecular Weight: 172,66 g/mol
Schmelzpunkt: 184 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00013381
CAS-Nummer: 60480-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR