2-Chlor-3-methylpyridin-N-oxid 98%
Lieferant: Apollo Scientific
OR902100-1G
OR902100-250MG
APOSOR902100-1GEA
364
EUR
APOSOR902100-1G
APOSOR902100-250MG
2-Chlor-3-methylpyridin-N-oxid 98%
2-Chlor-3-methylpyridin-N-oxid
Formel:
C₆H₆ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD14582951 CAS-Nummer: 91668-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...