2-(Cyclopropylamino)nicotinonitril 98%

Lieferant: Apollo Scientific

OR902105-1G OR902105-5G
APOSOR902105-1GEA 127 EUR
APOSOR902105-1G APOSOR902105-5G
2-(Cyclopropylamino)nicotinonitril 98%
2-(Cyclopropylamino)nicotinonitril
Formel: C₉H₉N₃
Molecular Weight: 159.190 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09455268
CAS-Nummer: 52583-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR