2-(Butylamino)nicotinonitrile 98%
Lieferant: Apollo Scientific
Synonyme:
3-Cyano-2-butylaminopyridine
OR902106-1G
OR902106-25G
OR902106-5G
APOSOR902106-1GEA
125
EUR
APOSOR902106-1G
APOSOR902106-25G
APOSOR902106-5G
2-(Butylamino)nicotinonitrile 98%
2-(Butylamino)nicotinonitrile
Formel:
C₁₀H₁₃N₃ Molecular Weight: 175,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08460989 CAS-Nummer: 74611-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...