5-Methoxypicolinamid 98%
Lieferant: Apollo Scientific
OR902143-1G
OR902143-250MG
APOSOR902143-1GEA
364
EUR
APOSOR902143-1G
APOSOR902143-250MG
5-Methoxypicolinamid 98%
5-Methoxypicolinamid
Formel:
C₇H₈N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD11976465 CAS-Nummer: 88166-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...