N-Phenylglycinonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 2-(Phenylamino)acetonitrile

OR902161-100G
APOSOR902161-100GEA 26.1 EUR
APOSOR902161-100G
N-Phenylglycinonitrile 98%
N-Phenylglycinonitrile
Formel: C₈H₈N₂
Molecular Weight: 132,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044430
CAS-Nummer: 3009-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR