6-Brom-o-toluonitril 95%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-6-methylbenzonitrile
OR902209-5G
APOSOR902209-5GEA
149
EUR
APOSOR902209-5G
6-Brom-o-toluonitril 95%
6-Brom-o-toluonitril
Formel:
C₈H₆BrN Molecular Weight: 196,0439 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09834775 CAS-Nummer: 77532-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...