3-Methoxy-2-nitrobenzaldehyd 95%
Lieferant: Apollo Scientific
OR902223-5G
APOSOR902223-5GEA
217
EUR
APOSOR902223-5G
3-Methoxy-2-nitrobenzaldehyd 95%
3-Methoxy-2-nitrobenzaldehyd
Formel:
C₈H₇NO₄ Molecular Weight: 181,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007135 CAS-Nummer: 53055-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...