Ethyl-6-(hydroxymethyl)pyridine-2-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 6-(hydroxymethyl)pyridine-2-carboxylate

OR902265-1G OR902265-5G
APOSOR902265-1GEA 183 EUR
APOSOR902265-1G APOSOR902265-5G
Ethyl-6-(hydroxymethyl)pyridine-2-carboxylate 98%
Ethyl-6-(hydroxymethyl)pyridine-2-carboxylate
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750009
CAS-Nummer: 41337-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR