7-Chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR902369-1G
OR902369-5G
APOSOR902369-1GEA
255
EUR
APOSOR902369-1G
APOSOR902369-5G
7-Chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine 98%
7-Chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine
Formel:
C₈H₈ClNO Molecular Weight: 169,6082 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07364075 CAS-Nummer: 113770-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...