1-Cyclopentylpiperidin-3-one 95%

Lieferant: Apollo Scientific

OR902395-100MG OR902395-1G OR902395-250MG
APOSOR902395-100MGEA 183 EUR
APOSOR902395-100MG APOSOR902395-1G APOSOR902395-250MG
1-Cyclopentylpiperidin-3-one 95%
1-Cyclopentylpiperidin-3-one
Formel: C₁₀H₁₇NO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 886365-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR