4-Chloropyrido[3,4-d]pyrimidine 95%
Lieferant: Apollo Scientific
OR902458-1G
OR902458-250MG
APOSOR902458-1GEA
249
EUR
APOSOR902458-1G
APOSOR902458-250MG
4-Chloropyrido[3,4-d]pyrimidine 95%
4-Chloropyrido[3,4-d]pyrimidine
Formel:
C₇H₄ClN₃ Molecular Weight: 165,5798 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09999184 CAS-Nummer: 51752-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...