1-tert-Butyl-5-oxopyrrolidine-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR902496-1G OR902496-250MG OR902496-5G
APOSOR902496-1GEA 219 EUR
APOSOR902496-1G APOSOR902496-250MG APOSOR902496-5G
1-tert-Butyl-5-oxopyrrolidine-3-carboxylic acid 95%
1-tert-Butyl-5-oxopyrrolidine-3-carboxylic acid
Formel: C₉H₁₅NO₃
Molecular Weight: 185.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04989713
CAS-Nummer: 773865-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR