6-Bromo-1,2-benzisoxazole 95%

Lieferant: Apollo Scientific

OR902552-1G
APOSOR902552-1GEA 115 EUR
APOSOR902552-1G
6-Bromo-1,2-benzisoxazole 95%
6-Bromo-1,2-benzisoxazole
Formel: C₇H₄BrNO
Molecular Weight: 198,0168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659634
CAS-Nummer: 1060802-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR