5-Bromo-4-chloropyrimidine 96%

Lieferant: Apollo Scientific

OR902595-5G
APOSOR902595-5GEA 88.6 EUR
APOSOR902595-5G
5-Bromo-4-chloropyrimidine 96%
5-Bromo-4-chloropyrimidine
Formel: C₄H₂BrClN₂
Molecular Weight: 193,4291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750158
CAS-Nummer: 56181-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR