Buthionine sulphoximine

Lieferant: Apollo Scientific
Achtung

Synonyme: DL-Buthionine (S,R)-sulphoximine

OR9025T-1G OR9025T-250MG
APOSOR9025T-1GEA 415 EUR
APOSOR9025T-1G APOSOR9025T-250MG
Buthionine sulphoximine
Buthionine sulphoximine

A potent & specific inhibitor of g-glutamylcysteine synthetase.

Formel: C₈H₁₈N₂O₃S
Molecular Weight: 222.303 g/mol
Schmelzpunkt: 214…215 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00070309
CAS-Nummer: 5072-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR