3,4-Dimethylpyridin-2-amine 95%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-3,4-dimethylpyridine

OR902657-1G OR902657-250MG
APOSOR902657-1GEA 246 EUR
APOSOR902657-1G APOSOR902657-250MG
3,4-Dimethylpyridin-2-amine 95%
3,4-Dimethylpyridin-2-amine
Formel: C7H10N2
Molecular Weight: 122,1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02247162
CAS-Nummer: 823-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR