4-(4-Hydroxyphenyl)butanoic acid 95%

Lieferant: Apollo Scientific

OR902708-10G OR902708-1G OR902708-5G
APOSOR902708-10GEA 220 EUR
APOSOR902708-10G APOSOR902708-1G APOSOR902708-5G
4-(4-Hydroxyphenyl)butanoic acid 95%
4-(4-Hydroxyphenyl)butanoic acid
Formel: C₁₀H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00033287
CAS-Nummer: 7021-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR