(R)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine 95%

Lieferant: Apollo Scientific

OR902781-1G OR902781-250MG
APOSOR902781-1GEA 132 EUR
APOSOR902781-1G APOSOR902781-250MG
(R)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine 95%
(R)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine
Formel: C₁₁H₂₂N₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 155726-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR