1-Quinolin-5-ylmethylamine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 1-Quinolin-5-yl-methylamine hydrochloride

OR902830-1G OR902830-250MG
APOSOR902830-1GEA 322 EUR
APOSOR902830-1G APOSOR902830-250MG
1-Quinolin-5-ylmethylamine hydrochloride 95%
1-Quinolin-5-ylmethylamine hydrochloride
Formel: C₁₀H₁₁ClN₂
Molecular Weight: 194,6600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09701405
CAS-Nummer: 1187931-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR