1-(3-Aminopropyl)pyrrolidin 97%

Lieferant: Apollo Scientific

Synonyme: N-(3-Aminopropyl)pyrrolidine, 3-(Pyrrolidin-1-yl)propan-1-amine

OR902892-100G OR902892-25G
APOSOR902892-100GEA 388 EUR
APOSOR902892-100G APOSOR902892-25G
1-(3-Aminopropyl)pyrrolidin 97%
1-(3-Aminopropyl)pyrrolidin
Formel: C₇H₁₆N₂
Molecular Weight: 128,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014100
CAS-Nummer: 23159-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR