Ethyl acetimidate hydrochloride 95%
Lieferant: Apollo Scientific
OR903066-100G OR903066-500G
APOSOR903066-100GEA 105 EUR
APOSOR903066-100G APOSOR903066-500G
Ethyl acetimidate hydrochloride 95%
Ethyl acetimidate hydrochloride
Formel: C₄H₁₀ClNO
Molecular Weight: 123.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012572
CAS-Nummer: 2208-07-3
Bestellen
TECHNISCHE DATEN
Environmentally Preferable