2,4-Dimethyl-3-nitropyridine

Lieferant: Apollo Scientific

OR9031-1G OR9031-250MG
APOSOR9031-1GEA 78.3 EUR
APOSOR9031-1G APOSOR9031-250MG
2,4-Dimethyl-3-nitropyridine
2,4-Dimethyl-3-nitropyridine
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08235269
CAS-Nummer: 1074-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR