3-(3-Hydroxyphenyl)propionsäure 97%

Lieferant: Apollo Scientific

Synonyme: 3-(3-Hydroxyphenyl)propanoic acid

OR903114-25G
APOSOR903114-25GEA 102 EUR
APOSOR903114-25G
3-(3-Hydroxyphenyl)propionsäure 97%
3-(3-Hydroxyphenyl)propionsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016554
CAS-Nummer: 621-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR