2-(3,4-Dimethylphenyl)pyrrolidine 95%

Lieferant: Apollo Scientific

OR903115-250MG
APOSOR903115-250MGEA 0 EUR
APOSOR903115-250MG
2-(3,4-Dimethylphenyl)pyrrolidine 95%
2-(3,4-Dimethylphenyl)pyrrolidine
Formel: C₁₂H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD05189449
CAS-Nummer: 881040-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR