2-Bromo-5-methylpyrimidine 98%

Lieferant: Apollo Scientific

OR903299-1G OR903299-5G
APOSOR903299-1GEA 270 EUR
APOSOR903299-1G APOSOR903299-5G
2-Bromo-5-methylpyrimidine 98%
2-Bromo-5-methylpyrimidine
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09753737
CAS-Nummer: 150010-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR