5-Bromo-6-chloropyrimidin-4-amine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR903308-5G
APOSOR903308-5GEA
85.7
EUR
APOSOR903308-5G
5-Bromo-6-chloropyrimidin-4-amine 98%
5-Bromo-6-chloropyrimidin-4-amine
Formel:
C₄H₃BrClN₃ Molecular Weight: 208,44 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09999224 CAS-Nummer: 663193-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...