2-(4-Isopropylphenoxy)acetic acid 95%

Lieferant: Apollo Scientific

OR903364-25G
APOSOR903364-25GEA 112 EUR
APOSOR903364-25G
2-(4-Isopropylphenoxy)acetic acid 95%
2-(4-Isopropylphenoxy)acetic acid
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014364
CAS-Nummer: 1643-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR