2-Amino-4-brom-6-methylpyrimidin 98%

Lieferant: Apollo Scientific

OR903436-1G OR903436-5G
APOSOR903436-1GEA 270 EUR
APOSOR903436-1G APOSOR903436-5G
2-Amino-4-brom-6-methylpyrimidin 98%
2-Amino-4-bromo-6-methylpyrimidine
Formel: C₅H₆BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11109799
CAS-Nummer: 5734-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR