4-Chloro-2-(methylsulphanyl)pyridine 98%
Lieferant: Apollo Scientific
OR903450-1G
OR903450-250MG
APOSOR903450-1GEA
415
EUR
APOSOR903450-1G
APOSOR903450-250MG
4-Chloro-2-(methylsulphanyl)pyridine 98%
4-Chloro-2-(methylsulphanyl)pyridine
Formel:
C₆H₆ClNS Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109868 CAS-Nummer: 334542-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...